missing translation for 'onlineSavingsMsg'
Learn More

Propargyl p-toluenesulfonate, 97%

Product Code. 15438949
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438949 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15438949 Supplier Thermo Scientific Alfa Aesar Supplier No. L12075.09
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6165-76-0 | C10H10O3S | 210.247 g/mol

Propargyl p-toluenesulfonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Propargyl p-toluenesulfonate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in Chloroform, Dichloromethane, DMSO, Ethyl Aceta
TRUSTED_SUSTAINABILITY

Specifications

CAS 6165-76-0
Density 1.22
Boiling Point 117°C to 121°C (0.3 mmHg)
Molecular Formula C10H10O3S
Refractive Index 1.527
MDL Number MFCD00078365
Quantity 10 g
Beilstein 1212957
Synonym propargyl p-toluenesulfonate, propargyl tosylate, prop-2-yn-1-yl 4-methylbenzenesulfonate, p-toluenesulfonic acid, 2-propynyl ester, 2-propyn-1-ol, 4-methylbenzenesulfonate, ccris 9146, p-toluenesulfonic acid propargyl ester, propargylp-toluenesulfonate, propargyl p-toluene sulfonate, acmc-209mv7
Solubility Information Soluble in Chloroform,Dichloromethane,DMSO,Ethyl Acetate. Not miscible in water.
InChI Key LMBVCSFXFFROTA-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCC#C
IUPAC Name prop-2-ynyl 4-methylbenzenesulfonate
Molecular Weight (g/mol) 210.247
PubChem CID 22547
Formula Weight 210.25
Percent Purity 97%
Chemical Name or Material Propargyl p-toluenesulfonate
Show More Show Less
compliance-icons
Product Identifier
  • Propargyl p-toluenesulfonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Propargyl p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.