missing translation for 'onlineSavingsMsg'
Learn More

Propargyl benzoate, 98%

Product Code. 15410268
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15410268

Brand: Thermo Scientific Alfa Aesar H53444.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6750-04-5 | C9H12N3Na2O8P | 367.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6750-04-5

Specifications

Density 1.106
Boiling Point 225°C to 226°C
Molecular Formula C10H8O2
Linear Formula C6H5CO2CH2C≡CH
Refractive Index 1.532
MDL Number MFCD00996341
Quantity 25 g
Synonym prop-2-yn-1-yl benzoate, propargyl benzoate, prop-2-yn-1-ylbenzoate, 2-propynyl benzoate, acmc-20ajtj, 2-propynyl benzoate #
InChI Key NBDHEMWCIUHARG-UHFFFAOYSA-N
SMILES C#CCOC(=O)C1=CC=CC=C1
IUPAC Name prop-2-ynyl benzoate
Molecular Weight (g/mol) 160.17
PubChem CID 336291
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material Propargyl benzoate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.