missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propargyl alcohol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
Propargyl alcohol is a solvent stabilizer that is useful as an intermediate in organic synthesis, an electroplating brightener additive. It is useful 3-carbon fragment in acetylene coupling reactions, It undergoes Pd-catalyzed O-coupling with alcohols to give the corresponding 1-methoxyallyl ethers.
Solubility
Fully miscible in water.
Notes
Store at 4°C. Incompatible with oxidizing agent, strong acids, strong bases, amines, phosphorous pentoxide, isocyanate vapor & heat.
Specifications
Melting Point | -53°C |
Density | 0.949 |
Boiling Point | 114°C to 115°C |
Flash Point | 33°C (91°F) |
Linear Formula | HC≡CCH2OH |
Refractive Index | 1.432 |
Quantity | 2500 mL |
UN Number | UN2929 |
Beilstein | 506003 |
Merck Index | 14,7809 |
Mostrar más |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'