missing translation for 'onlineSavingsMsg'
Learn More

Propargyl acetate, 97%

Product Code. 11342407
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11342407 25 g 25g
11332407 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11342407 Supplier Thermo Scientific Alfa Aesar Supplier No. L10031.14
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 627-09-8 | C5H6O2 | 98.101 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-09-8
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00041601
InChI Key RIZZXCJMFIGMON-UHFFFAOYSA-N
Synonym propargyl acetate, 2-propyn-1-ol, acetate, 3-acetoxypropyne, 2-propynyl acetate, propargyl alcohol acetate, prop-2-yn-1-yl acetate, acmc-1awj3, 2-propyn-1-ol, 1-acetate, 4-02-00-00197 beilstein handbook reference
PubChem CID 69388
IUPAC Name prop-2-ynyl acetate
SMILES CC(=O)OCC#C

Specifications

Density 0.989
Boiling Point 124°C
Flash Point 31°C (88°F)
Odor Characteristic
Linear Formula CH3CO2CH2C≡CH
Refractive Index 1.4185
Quantity 25 g
UN Number UN1992
Beilstein 1742046
Formula Weight 98.1
Percent Purity 97%
Chemical Name or Material Propargyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.