missing translation for 'onlineSavingsMsg'
Learn More

Propane-1,3-diylbis((3-acetamidopropyl)carbamic Acid) di-tert-Butyl Ester-d6, TRC

Product Code. 30421389
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30421389 1 mg 1mg
30409704 2.5 mg 2.5mg
30420070 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30421389 Supplier Toronto Research Chemicals Supplier No. TRCP8352871MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Propane-1,3-diylbis((3-acetamidopropyl)carbamic Acid) di-tert-Butyl Ester-d6
Molecular Formula C23 D6 H38 N4 O6
Quantity 1 mg
InChI Formula InChI=1S/C23H44N4O6/c1-18(28)24-12-9-14-26(20(30)32-22(3,4)5)16-11-17-27(15-10-13-25-19(2)29)21(31)33-23(6,7)8/h9-17H2,1-8H3,(H,24,28)(H,25,29)/i1D3,2D3
SMILES [2H]C([2H])([2H])C(=O)NCCCN(CCCN(CCCNC(=O)C([2H])([2H])[2H])C(=O)OC(C)(C)C)C(=O)OC(C)(C)C
Molecular Weight (g/mol) 478.656
Formula Weight 478.364
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.