missing translation for 'onlineSavingsMsg'
Learn More

Propane-1,3-Diol, Pure, Fisher Chemical

Product Code. 10244060 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10244060 500 mL Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10244060 Supplier Fisher Chemical Supplier No. P/7450/08

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 504-63-2
Melting Point -32°C
Color Colorless
pH 4.5 to 7.0
Boiling Point 214°C
Packaging Amber glass bottle
Molecular Formula C3H8O2
MDL Number MFCD00002949
Quantity 500 mL
Synonym 1,3-propanediol, trimethylene glycol, 1,3-dihydroxypropane, 2-deoxyglycerol, 1,3-propylene glycol, 1,3-propylenediol, 1,3-propandiol, 2-hydroxymethyl ethanol, omega-propanediol, beta-propylene glycol
InChI Key YPFDHNVEDLHUCE-UHFFFAOYSA-N
SMILES OCCCO
IUPAC Name propane-1,3-diol
Molecular Weight (g/mol) 76.10
PubChem CID 10442
ChEBI CHEBI:16109
Formula Weight 76.09g/mol
Vapor Pressure <0.1mbar at 20°C
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.