missing translation for 'onlineSavingsMsg'
Learn More

PropamocarbHydrochloride, TRC

Product Code. 30012266
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012266 10 g 10g
30011892 25 g 25g
30005895 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012266 Supplier Toronto Research Chemicals Supplier No. TRCP01463610G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25606-41-1
Molecular Formula C9 H20 N2 O2 . Cl H
Molecular Weight (g/mol) 224.73
Synonym Propamocarb hydrochloride, Carbamic acid, N-[3-(dimethylamino)propyl]-, propyl ester, hydrochloride (1:1) (ACI), Carbamic acid, [3-(dimethylamino)propyl]-, propyl ester, monohydrochloride (8CI, 9CI), Gentracur, Previcur Flex, Previcur N, Propamocarb hydrochloride, Propyl 3-(dimethylamino)propylcarbamate monohydrochloride, SN 66752
IUPAC Name propyl N-[3-(dimethylamino)propyl]carbamate;hydrochloride
SMILES Cl.CCCOC(=O)NCCCN(C)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H20N2O2.ClH/c1-4-8-13-9(12)10-6-5-7-11(2)3;/h4-8H2,1-3H3,(H,10,12);1H
Formula Weight 224.1292
Chemical Name or Material Propamocarb hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.