missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ ProPak™ Recombinant Human IL-2 GMP Protein, CF
Click to view available options
Quantity:
150,000 IU
3,000,000 IU
Unit Size:
Each
Description
Measured in a cell proliferation assay using CTLL-2 mouse cytotoxic T cells. Gearing, A.J.H. and C.B. Bird (1987) in Lymphokines and Interferons, A Practical Approach. Clemens, M.J. et al. (eds): IRL Press. 295. The potency of recombinant human IL-2 is 75-125% of the IU pack size. International units are calibrated from an internal reference standard that is value assigned against the human IL-2 WHO International Standard (NIBSC code: 86/500).
Specifications
Specifications
| Accession Number | P60568.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Supplied as a 0.2 μm filtered solution in PBS, recombinant HSA, and Trehalose. |
| Gene ID (Entrez) | 3558 |
| Molecular Weight (g/mol) | M.W.-observed: 13 kDa, under reducing conditions., M.W.-theroretical: 15.5 kDa |
| Quantity | 150,000 IU |
| Source | E. coli-derived human IL-2 protein Ala21-Thr153 (Cys145Ser), with and without an N-terminal Met Produced using non-animal reagents in an animal-free laboratory. Manufactured and tested under cGMP guidlelines. |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. A minimum of 6 months when stored between -14 °C and -40 °C. Can be stored up to 2 weeks at 2-8 °C. Refer to lot specific COA for the Use by Date. |
| Endotoxin Concentration | <5.0 EU/mL by the LAL method. |
| Gene Alias | aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction