missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propafenone hydrochloride, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Propafenone (hydrochloride) (SA-79 (hydrochloride)) is a class of anti-arrhythmic medication, which treats illnesses associated with rapid heart beats such as atrial and ventricular arrhythmias.
Specifications
Specifications
| Chemical Name or Material | Propafenone hydrochloride |
| CAS | 34183-22-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H28ClNO3 |
| Quantity | 1 g |
| Synonym | SA-79 hydrochloride |
| Solubility Information | DMSO : 33.33 mg/mL (88.20 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.77 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC=CC=C1OCC(O)CNCCC)CCC2=CC=CC=C2.[H]Cl |
| Molecular Weight (g/mol) | 377.9 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction