missing translation for 'onlineSavingsMsg'
Learn More

Propafenone hydrochloride, MedChemExpress

Product Code. 16430928
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16430928

Brand: Medchem Express HYB0432A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Propafenone (hydrochloride) (SA-79 (hydrochloride)) is a class of anti-arrhythmic medication, which treats illnesses associated with rapid heart beats such as atrial and ventricular arrhythmias.

Propafenone (hydrochloride) (SA-79 (hydrochloride)) is a class of anti-arrhythmic medication, which treats illnesses associated with rapid heart beats such as atrial and ventricular arrhythmias.

Specifications

Chemical Name or Material Propafenone hydrochloride
CAS 34183-22-7
Color White
Purity Grade Notes Research
Molecular Formula C21H28ClNO3
Quantity 5 g
Synonym SA-79 hydrochloride
Solubility Information DMSO : 33.33 mg/mL (88.20 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.77 mM; Need ultrasonic)
SMILES O=C(C1=CC=CC=C1OCC(O)CNCCC)CCC2=CC=CC=C2.[H]Cl
Molecular Weight (g/mol) 377.9
Formula Weight 377.9
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.