missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propafenone EP Impurity F, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Propafenone EP Impurity F
Specifications
Specifications
| Chemical Name or Material | 1,1'-[2-Hydroxypropane-1,3-diyl-bis(oxy-2,1-phenylene)]bis(3-phenylpropan-1-one) |
| CAS | 1329643-40-4 |
| Analyte or Component Names | 1,1'-[2-Hydroxypropane-1,3-diyl-bis(oxy-2,1-phenylene)]bis(3-phenylpropan-1-one) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C33 H32 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Propafenone Hydrochloride Imp. F (EP), Propafenone Imp. F (EP), 1,1'-[2-Hydroxypropane-1,3-diyl-bis(oxy-2,1-phenylene)]bis(3-phenylpropan-1-one), Propafenone Hydrochloride Impurity F, Propafenone Impurity F, 1,1'-[Hydroxypropane-1,3-diylbis(oxy-2,1-phenylene)]bis(3-phenylpropan-1-one) |
| InChI Formula | InChI=1S/C25H31NO3.C2H2O4/c1-4-7-11-23-24(21-10-8-9-12-22(21)29-23)25(27)19-13-15-20(16-14-19)28-18-17-26(5-2)6-3;3-1(4)2(5)6/h8-10,12-16H,4-7,11,17-18H2,1-3H3;(H,3,4)(H,5,6) |
| SMILES | OC(COc1ccccc1C(=O)CCc2ccccc2)COc3ccccc3C(=O)CCc4ccccc4 |
| IUPAC Name | 1-[2-[2-hydroxy-3-[2-(3-phenylpropanoyl)phenoxy]propoxy]phenyl]-3-phenylpropan-1-one |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction