missing translation for 'onlineSavingsMsg'
Learn More

Propafenone EP Impurity C, Mikromol™

Product Code. 17932306
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17932306 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932306 Supplier Mikromol Supplier No. MM0405.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Propafenone EP Impurity C
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-[2-[((2RS)-Oxiranyl)methoxy]phenyl]-3-phenylpropan-1-one
CAS 22525-95-7
Analyte or Component Names 1-[2-[((2RS)-Oxiranyl)methoxy]phenyl]-3-phenylpropan-1-one
Shipping Condition Ice pack (-20°C)
Molecular Formula C18 H18 O3
Certifications/Compliance ISO 17025
Synonym 1-[2-[[(2RS)-Oxiranyl]methoxy]phenyl]-3-phenylpropan-1-one, Propafenone Hydrochloride Imp. C (EP), Propafenone Imp. C (EP), 1-[2-[[(2RS)-Oxiranyl]methoxy]phenyl]-3-phenylpropan-1-one, Propafenone Hydrochloride Impurity C, Propafenone Impurity C
InChI Formula InChI=1S/C18H18O3/c19-17(11-10-14-6-2-1-3-7-14)16-8-4-5-9-18(16)21-13-15-12-20-15/h1-9,15H,10-13H2
SMILES O=C(CCc1ccccc1)c2ccccc2OCC3CO3
IUPAC Name 1-[2-(oxiran-2-ylmethoxy)phenyl]-3-phenylpropan-1-one
Molecular Weight (g/mol) 282.33
Quantity 25 mg
Formula Weight 282.1256 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.