Learn More
MP Biomedicals™ Isis™ DNA Polymerase
A proofreading DNA polymerase and amplifies DNA with 40 times more accuracy than standard Taq DNA polymerase
Brand:  MP Biomedicals™ 11EPSIS100
Code : 80
Additional Details : Weight : 0.10000kg
Product Code. 11407010
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • Ideal for high-fidelity amplification
  • Use to amplify DNA with difficult to read secondary structures
  • Optimal for blunt-end PCR cloning
  • The most thermostable polymerase available
  • Useful to amplify GC rich sequences
  • Free of RNases and endo- and exonucleases
  • Research Use Only (RUO)
Specifications
Specifications
1 U/μL
100 U