missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prominin 2 Antibody (OTI6B7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73616JF549
This item is not returnable.
View return policy
Description
Prominin 2 Monoclonal specifically detects Prominin 2 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Prominin 2 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PROM2 | |
| Full-length protein expressed in 293T cell transfected with human Prominin 2 expression vector. | |
| 0.1 mL | |
| 150696.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI6B7 | |
| Western Blot, Immunohistochemistry | |
| hPROML2, MGC138714, PROM-2, prominin 2, prominin-2, Prominin-like protein 2, prominin-related protein, PROML2 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction