missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prometon-d14 (di-iso-propyl-d14), CDN
Specifications
Specifications
| Chemical Name or Material | Prometon D14 (di-iso-propyl D14) |
| CAS | 1219803-45-8 |
| Molecular Formula | C10 2H14 H5 N5 O |
| Quantity | 0.01 g |
| Synonym | Prometon D14 (di-iso-propyl D14) |
| InChI Formula | InChI=1 S/C10H19N5O/c1-6(2)11-8-13-9(12-7(3)4)15-10(14-8)16-5/h6-7 H,1-5H3,(H2,11,12,13,14,15)/i1D3,2D3,3D3,4D3,6 D,7 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(Nc1nc(NC([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H])nc(OC)n1)C([2 H])([2 H])[2 H] |
| IUPAC Name | 2-N,4-N-bis(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-6-methoxy-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 239.38 |
| Formula Weight | 239.2468 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?