missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Sulphoxide N-Oxide Hydrochloride, Mikromol™

Product Code. 30049685
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30049685 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049685 Supplier Mikromol Supplier No. MM0412.05

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Promethazine Sulphoxide N-Oxide Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promethazine Sulphoxide N-Oxide Hydrochloride
CAS 1135334-50-7
Analyte or Component Names Promethazine Sulphoxide N-Oxide Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C17 H20 N2 O2 S . Cl H
Certifications/Compliance ISO 9001
Synonym Promethazine Sulphoxide N-Oxide Hydrochloride
InChI Formula InChI=1S/C17H20N2O2S.ClH/c1-13(19(2,3)20)12-18-14-8-4-6-10-16(14)22(21)17-11-7-5-9-15(17)18;/h4-11,13H,12H2,1-3H3;1H
SMILES Cl.CC(CN1c2ccccc2S(=O)c3ccccc13)[N+](C)(C)[O-]
IUPAC Name N,N-dimethyl-1-(5-oxophenothiazin-10-yl)propan-2-amine oxide;hydrochloride
Molecular Weight (g/mol) 352.88
Quantity 100 mg
Formula Weight 352.1012 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.