missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Hydrochloride, TRC

Product Code. 30422738
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422738 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422738 Supplier Toronto Research Chemicals Supplier No. TRCP7570001G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promethazine Hydrochloride
CAS 58-33-3
Molecular Formula C17 H20 N2 S . Cl H
Quantity 1 g
Synonym Promethazine Hydrochloride, 10H-Phenothiazine-10-ethanamine','N,N,α-trimethyl-','hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine','N,N,α-trimethyl-','monohydrochloride (9CI), Phenothiazine','10-[2-(dimethylamino)propyl]-','monohydrochloride (8CI), 10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride, Anergan 25, Anergan 50, Atosil, Diprasine, Diprazin, Dorme, Duplamin, Fellozine, Fenazil, Fenergan, Genphen, Hiberna, Himazin, Kinetosin, Lergigan, Mepergan, NSC 231688, Phanergan D, Phena, Phencen, Phenergan, Phenergan VC, Pipolfen, Pipolphen, Plletia, Promasun, Promethazine chloride, Promethazine hydrochloride, Promethazine monohydrochloride, Promethazinium chloride, Prorex, Prothazine, Provigan, Pyrethia, Remsed, Romergan, Romergan hydrochloride
InChI Formula InChI=1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H
SMILES Cl.CC(CN1c2ccccc2Sc3ccccc13)N(C)C
IUPAC Name N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
Molecular Weight (g/mol) 320.88
Formula Weight 320.1114
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.