missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Hydrochloride, TRC

Product Code. 30422513
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30422513 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30422513 Supplier Toronto Research Chemicals Supplier No. TRCP757000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promethazine Hydrochloride
CAS 58-33-3
Molecular Formula C17 H20 N2 S . Cl H
Quantity 100 mg
Synonym Promethazine Hydrochloride, 10H-Phenothiazine-10-ethanamine','N,N,α-trimethyl-','hydrochloride (1:1), 10H-Phenothiazine-10-ethanamine','N,N,α-trimethyl-','monohydrochloride (9CI), Phenothiazine','10-[2-(dimethylamino)propyl]-','monohydrochloride (8CI), 10-[2-(Dimethylamino)propyl]phenothiazine hydrochloride, Anergan 25, Anergan 50, Atosil, Diprasine, Diprazin, Dorme, Duplamin, Fellozine, Fenazil, Fenergan, Genphen, Hiberna, Himazin, Kinetosin, Lergigan, Mepergan, NSC 231688, Phanergan D, Phena, Phencen, Phenergan, Phenergan VC, Pipolfen, Pipolphen, Plletia, Promasun, Promethazine chloride, Promethazine hydrochloride, Promethazine monohydrochloride, Promethazinium chloride, Prorex, Prothazine, Provigan, Pyrethia, Remsed, Romergan, Romergan hydrochloride
InChI Formula InChI=1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H
SMILES Cl.CC(CN1c2ccccc2Sc3ccccc13)N(C)C
IUPAC Name N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
Molecular Weight (g/mol) 320.88
Formula Weight 320.1114
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.