missing translation for 'onlineSavingsMsg'
Learn More

Promethazine Hydrochloride 1.0 mg/ml in Dimethyl Sulfoxide (as free base), LoGiCal

Product Code. 30462294
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462294 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462294 Supplier LoGiCal Supplier No. LGCAMP0412.0029

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Promethazine Hydrochloride 1.0 mg/ml in Dimethyl Sulfoxide (as free base)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promethazine Hydrochloride
CAS 58-33-3
Analyte or Component Names Promethazine Hydrochloride
Shipping Condition Ice Pack (-20°C)
Molecular Formula C17 H20 N2 S . Cl H
Certifications/Compliance ISO 17034
Synonym Promethazine Hydrochloride
InChI Formula InChI=1 S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13 H,12H2,1-3H3;1 H
SMILES Cl.CC(CN1c2ccccc2Sc3ccccc13)N(C)C
IUPAC Name N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Forensics
Molecular Weight (g/mol) 320.88
Quantity 1 mL
Formula Weight 320.1114 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.