missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Promethazine.HCl, 1mg/ml in Dmf (as free base), Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Promethazine.HCl, 1mg/ml in Dmf (as free base)
Specifications
Specifications
| Chemical Name or Material | Promethazine Hydrochloride |
| CAS | 58-33-3 |
| Analyte or Component Names | Promethazine Hydrochloride |
| Shipping Condition | Ice Pk. (-20°C) |
| Molecular Formula | C17 H20 N2 S . Cl H |
| Concentration | 1 mg/mL |
| Synonym | Promethazine Hydrochloride |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Cl.CC(CN1c2ccccc2Sc3ccccc13)N(C)C |
| IUPAC Name | N,N-dimethyl-1-phenothiazin-10-ylpropan-2-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction