missing translation for 'onlineSavingsMsg'
Learn More

Promethazine-D3 Hydrochloride 0.1 mg/ml in Dimethyl Sulfoxide (as free base), LoGiCal

Product Code. 30463823
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30463823 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30463823 Supplier LoGiCal Supplier No. LGCAMP0412.8031
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promethazine-D3 HCl
CAS 1435933-86-0
Solvent or Matrix Dimethyl Sulfoxide
Molecular Formula C17 2H3 H17 N2 S . Cl H
Quantity 1 mL
Synonym Promethazine-D3 Hydrochloride, N,α-dimethyl-N-(methyl-d3)-10H-phenothiazine-10-ethanamine, monohydrochloride
InChI Formula InChI=1S/C17H20N2S.ClH/c1-13(18(2)3)12-19-14-8-4-6-10-16(14)20-17-11-7-5-9-15(17)19;/h4-11,13H,12H2,1-3H3;1H/i2D3;
SMILES Cl.[2H]C([2H])([2H])N(C)C(C)CN1c2ccccc2Sc3ccccc13
IUPAC Name N-methyl-1-phenothiazin-10-yl-N-(trideuteriomethyl)propan-2-amine;hydrochloride
Molecular Weight (g/mol) 323.9
Formula Weight 323.1302 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.