missing translation for 'onlineSavingsMsg'
Learn More

Promazine Sulphoxide 1.0 mg/ml in Methanol, LoGiCal

Product Code. 30462632
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 30462632

Brand: LoGiCal LGCAMP0290.0101

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Promazine Sulphoxide 1.0 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promazine Sulphoxide
CAS 146-21-4
Analyte or Component Names Promazine Sulphoxide
Shipping Condition Room Temperature
Molecular Formula C17 H20 N2 O S
Certifications/Compliance ISO 17034
Synonym 3-(10 H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide (Promazine Sulphoxide), Promazine Hydrochloride Imp. A (EP), Promazine Imp. A (EP), 3-(10 H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide, Promazine Sulphoxide, Promazine Hydrochloride Impurity A, Promazine Impurity A
InChI Formula InChI=1 S/C17H20N2OS/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)21(20)17-11-6-4-9-15(17)19/h3-6,8-11 H,7,12-13H2,1-2H3
SMILES CN(C)CCCN1c2ccccc2S(=O)c3ccccc13
IUPAC Name N,N-dimethyl-3-(5-oxophenothiazin-10-yl)propan-1-amine
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 300.42
Quantity 1 mL
Formula Weight 300.1296 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.