missing translation for 'onlineSavingsMsg'
Learn More

Promazine EP Impurity A, Mikromol™

Product Code. 30051826
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30051826

Brand: Mikromol MM0290.010025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Promazine EP Impurity A
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Promazine Sulphoxide
CAS 146-21-4
Analyte or Component Names Promazine Sulphoxide
Shipping Condition Room Temperature
Molecular Formula C17 H20 N2 O S
Certifications/Compliance ISO 17025
Synonym 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide (Promazine Sulphoxide), Promazine Hydrochloride Imp. A (EP), Promazine Imp. A (EP), 3-(10H-Phenothiazin-10-yl)-N,N-dimethylpropan-1-amine S-Oxide, Promazine Sulphoxide, Promazine Hydrochloride Impurity A, Promazine Impurity A
InChI Formula InChI=1S/C17H20N2OS/c1-18(2)12-7-13-19-14-8-3-5-10-16(14)21(20)17-11-6-4-9-15(17)19/h3-6,8-11H,7,12-13H2,1-2H3
SMILES CN(C)CCCN1c2ccccc2S(=O)c3ccccc13
IUPAC Name N,N-dimethyl-3-(5-oxophenothiazin-10-yl)propan-1-amine
Molecular Weight (g/mol) 300.42
Quantity 25 mg
Formula Weight 300.1296 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.