missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolyl endopeptidase-like Antibody (JE65-64), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32853
This item is not returnable.
View return policy
Description
Prolyl endopeptidase-like Monoclonal antibody specifically detects Prolyl endopeptidase-like in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| Prolyl endopeptidase-like | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, Immunohistochemistry, Immunohistochemistry-Paraffin 1:400 | |
| EC 3.4.21, EC 3.4.21.-, EC 3.4.21.83, KIAA0436FLJ16627, prolyl endopeptidase-like, Prolylendopeptidase-like, putative prolyl oligopeptidase | |
| Recombinant protein within human Prolyl endopeptidase-like aa 621-720/727. (Uniprot: Q4J6C6) | |
| 100 μL | |
| Cell Biology | |
| 9581 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| JE65-64 | |
| Unconjugated | |
| 1*TBS (pH7.4), 0.05% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction