missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin R Antibody (BAY 1158061) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-27967JF669
This item is not returnable.
View return policy
Description
Prolactin R Monoclonal antibody specifically detects Prolactin R in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Prolactin R | |
| Monoclonal | |
| Janelia Fluor 669 | |
| delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein | |
| PRLR/ Prolactin Receptor | |
| 0.1 mL | |
| Apoptosis | |
| 5618 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| BAY 1158061 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction