missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Prolactin R Monoclonal antibody specifically detects Prolactin R in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Specifications
Specifications
| Antigen | Prolactin R |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF |
| Classification | Monoclonal |
| Clone | B6.2 + PRLR742 |
| Conjugate | PerCP |
| Formulation | PBS |
| Gene Alias | delta 4-delta 7/11 truncated prolactin receptor, delta 4-SF1b truncated prolactin receptor, hPRL receptor, hPRLrI, PRL-R, prolactin receptor, prolactin receptor delta 7/11, secreted prolactin binding protein |
| Host Species | Mouse |
| Immunogen | Semi-Purified human Prolactin R (B6.2); Recombinant human Prolactin R (PRLR742) (Uniprot: P16471) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?