missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prolactin / PRL Mouse, Clone: PRL-Q7, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio QDX44-100ug
This item is not returnable.
View return policy
Specifications
| Prolactin / PRL | |
| Monoclonal | |
| Unconjugated | |
| NP_001157030 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
| ELISA | |
| PRL-Q7 | |
| PBS (pH 7.4) | |
| PRL | |
| Protein G | |
| RUO | |
| 5617 | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction