missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prolactin Monoclonal Antibody (P5C8A4)

Description
MIP0202 targets Prolactin in ELISA, IP, and RIA applications and shows reactivity with Human samples. The MIP0202 immunogen is human Prolactin. MIP0202 detects Prolactin which has a predicted molecular weight of approximately 25 kDa. MIP0202 was formerly sold as a Seradyn product.
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.
Specifications
Specifications
| Antigen | Prolactin |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | P5C8A4 |
| Concentration | 5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | PRL |
| Gene Accession No. | P01236 |
| Gene Alias | AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?