missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prolactin Monoclonal Antibody (C13), Invitrogen™

Mouse Monoclonal Antibody
£115.00 - £385.00
Specifications
| Antigen | Prolactin |
|---|---|
| Clone | C13 |
| Concentration | 1 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17654166
|
Invitrogen™
MA543711 |
20 μL |
£115.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17644166
![]() |
Invitrogen™
MA543710 |
100 μL |
£385.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Prolactin (PRL) is a pituitary hormone involved in the stimulation of milk production, salt and water regulation, growth, development and reproduction. PRL exerts its influence on different cell types through a signal transduction pathway which begins with the binding of the hormone to a transmembrane PRL receptor.Specifications
| Prolactin | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| AV290867; decidual prolactin; GHA1; growth hormone A1; Human prolactin; lactogenic prehormone (AA -30 to 199); preprolactin; pre-prolactin; Prl; Prl1a1; PRLB; PRLSD1; Prol; Prolactin; prolactin family 1, subfamily a, member 1; prolactin precursor; prolactin protein; RATPRLSD1; RNPROL | |
| PRL | |
| IgG2b κ | |
| Protein A/G | |
| Antibody |
| C13 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Rat | |
| P01236, P01237 | |
| 24683, 5617 | |
| Leu29-Cys227 (Accession P01236), with N-terminal His Tag. | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PRL |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title