missing translation for 'onlineSavingsMsg'
Learn More

Prolactin Antibody (1085531) [DyLight 594], Novus Biologicals™

Product Code. 30761901 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761901 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761901 Supplier Novus Biologicals Supplier No. FAB11590M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prolactin Monoclonal antibody specifically detects Prolactin in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prolactin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 1085531
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias prolactin
Host Species Mouse
Immunogen E. coli- derived human Prolactin, Leu29-Cys227, Accession # P01236
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Phospho Specific, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5617
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.