missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PROL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564238
This item is not returnable.
View return policy
Description
Immunogen sequence: ISSSTVPISS TPEPATSISA ATPAASTENT TQILANRPHT VLLNATVQVT TSNQTILSSP A Highest antigen sequence identity to the following orthologs: Mouse - 36%, Rat - 34%.
Specifications
| PROL1 | |
| Polyclonal | |
| Unconjugated | |
| OPRPN | |
| Basic proline-rich lacrimal protein; basic proline-rich protein; BPLP; opiorphin; Opiorphin prepropeptide; OPRPN; PRL1; PROL1; proline rich, lacrimal 1; proline-rich 1; proline-rich protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 58503 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q99935 | |
| OPRPN | |
| Recombinant protein corresponding to Human PROL1. Recombinant protein control fragment (Product #RP-103608). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction