missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PROL1 Polyclonal Antibody
GREENER_CHOICE

Goat Polyclonal Antibody

Brand:  Invitrogen™ PA5142656

Product Code. 17858622

  • £406.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

PROL1
Polyclonal
Unconjugated
Basic proline-rich lacrimal protein; basic proline-rich protein; BPLP; opiorphin; Opiorphin prepropeptide; OPRPN; PRL1; PROL1; proline rich, lacrimal 1; proline-rich 1; proline-rich protein 1
Peptide with sequence C-QFPVRKYLEDPRY.
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Goat
Ammonium sulfate precipitation
RUO
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.