missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Prokineticin Receptor 1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5102241
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PKR1.
GPR73A is a receptor for prokineticin 1. It is a member of the GPCR family and is coupled to the G(q) subclass of heteromeric G proteins. Activation of the receptor leads to mobilization of calcium, stimulation of phosphoinositide turnover and activation of p44/p42 mitogen-activated protein kinase. GPR73A expression has been reported in adrenal, peripheral blood leukocytes, brain, colon, pancreas, prostate, rectum, salivary gland, small intestine, spleen, stomach, testis, and thyroid. No human ESTs have been identified.
Specifications
| Prokineticin Receptor 1 | |
| Polyclonal | |
| Unconjugated | |
| PROKR1 | |
| EG-VEGF receptor; EG-VEGFR1; G protein-coupled receptor 73; G protein-coupled receptor ZAQ; GPCR73A; GPR73; GPR73a; G-protein coupled receptor 73; G-protein coupled receptor ZAQ; PKR1; PK-R1; Prokineticin receptor 1; Prokr1; ZAQ | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10887, 192648, 58182 | |
| -20°C | |
| Liquid |
| Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8R416, Q8TCW9, Q9JKL1 | |
| PROKR1 | |
| A synthesized peptide derived from human PROKR1(Accession Q8TCW9), corresponding to amino acid residues H26-C76. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction