missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prokineticin R1/PROKR1 Antibody (420849), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4655L
This item is not returnable.
View return policy
Description
Prokineticin R1/PROKR1 Monoclonal antibody specifically detects Prokineticin R1/PROKR1 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Prokineticin R1/PROKR1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 420849 | |
| Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 73, G protein-coupled receptor ZAQ, GPR73, GPR73aG-protein coupled receptor 73, PK-R1, PKR1G-protein coupled receptor ZAQ, prokineticin receptor 1, ZAQ | |
| NS0 mouse myeloma cell line transfected with human Prokineticin R1/PKR1, Met1-Lys393, Accession # Q8TCW9 | |
| 0.1 mL | |
| GPCR | |
| 10887 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction