missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Prokineticin 2 Polyclonal antibody specifically detects Prokineticin 2 in Human,Mouse samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | Prokineticin 2 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | BV8Protein Bv8 homolog, KAL4, MIT1, PK2prokineticin-2, prokineticin 2 |
| Host Species | Rabbit |
| Immunogen | prokineticin 2. (Uniprot# Q9HC23) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?