missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PROK1 Recombinant Rabbit Monoclonal Antibody (1B4G2)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA542698
This item is not returnable.
View return policy
Description
Positive test controls include: 293T, HepG2, Mouse liver, Mouse kidney, Rat ovary, Rat kidney. Immunogen sequence: MRGATRVSIM LLLVTVSDCA VITGACERDV QCGAGTCCAI SLWLRGLRMC TPLGREGEEC HPGSHKVPFF RKRKHHTCPC LPNLLCSRFP DGRYRCSMDL KNINF.
Endocrine gland-derived vascular endothelial growth factor induces proliferation, migration, and fenestration in capillary endothelial cells derived from endocrine glands. Its expression is induced by hypoxia and is restricted to the steroidogenic glands. Its expression is often complementary to the expression of VEGF, suggesting that these molecules function in a coordinated manner.
Specifications
| PROK1 | |
| Recombinant Monoclonal | |
| 0.33 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P58294, Q14A28, Q8R414 | |
| PROK1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-105 of human PROK1 (P58294). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot, Immunocytochemistry | |
| 1B4G2 | |
| Unconjugated | |
| PROK1 | |
| black mamba toxin-related protein; EGVEGF; EG-VEGF; endocrine-gland-derived vascular endothelial growth factor; Mambakine; PK1; PRK1; PROK1; prokineticin 1; prokineticin-1; UNQ600/PRO1186 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 192205, 246691, 84432 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction