missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proinsulin Antibody (C-PEP-01), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-413IR
This item is not returnable.
View return policy
Description
Proinsulin Monoclonal specifically detects Proinsulin in Human samples. It is validated for ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Proinsulin | |
| Monoclonal | |
| DyLight 755 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| The antibody C-PEP-01 reacts specifically with C-peptide, a part of the Proinsulin molecule. Proinsulin consists of the three parts: C-peptide and two long strands of amino acids (alpha and beta chains; later become linked together to form the Insulin molecule). No cross-reactivity with Insulin or other peptide hormones or proteins was observed. | |
| Store at 4C in the dark. |
| ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| C-PEP-01 | |
| ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| INS | |
| C-peptide (Human) conjugated to BSA. | |
| 0.1 mL | |
| Diabetes Research, Immune System Diseases, Immunology | |
| 3630 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction