missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prohibitin Mouse anti-Human, Clone: PHB/3226, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | Prohibitin |
|---|---|
| Clone | PHB/3226 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Western Blot, Immunohistochemistry (Paraffin), Peptide Array |
Description
Prohibitin Monoclonal specifically detects Prohibitin in Human samples. It is validated for Western Blot, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Prohibitin | |
| 0.2 mg/ml | |
| Western Blot, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 5245 | |
| IgG2b κ | |
| Protein A or G purified |
| PHB/3226 | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| PHB1, prohibitin | |
| Recombinant human Prohibitin protein fragment (aa167-261) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title