missing translation for 'onlineSavingsMsg'
Learn More

Prohibitin 2 Antibody (2D3), Novus Biologicals™

Product Code. 18376259 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18376259 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18376259 Supplier Novus Biologicals Supplier No. H00011331M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Prohibitin 2 Monoclonal antibody specifically detects Prohibitin 2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Prohibitin 2
Applications ELISA
Classification Monoclonal
Clone 2D3
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH14766
Gene Alias BAP, Bap37, BCAP37, B-cell associated protein, D-prohibitin, MGC117268, p22, PNAS-141, prohibitin 2, prohibitin-2, REAB-cell receptor-associated protein BAP37, Repressor of estrogen receptor activity
Host Species Mouse
Immunogen PHB2 (AAH14766, 37 a.a. ~ 299 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RESVFTVEGGHRAIFFNRIGGVQQDTILAEGLHFRIPWFQYPIIYDIRARPRKISSPTGSKDLQMVNISLRVLSRPNAQELPSMYQRLGLDYEERVLPSIVNEVLKSVVAKFNASQLITQRAQVSLLIRRELTERAKDFSLILDDVAITELSFSREYTAAVEAKQVAQQEAQRAQFLVEKAKQEQRQKIVQAEGEAEAAKMLGEALSKNPGYIKLRKIRAAQNISKTIATSQNRIYLTADNLVLNLQDESFTRGSDSLIKGKK
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11331
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.