missing translation for 'onlineSavingsMsg'
Learn More

Proguanil, MedChemExpress

Product Code. 16434497
Encompass_Preferred
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
This item is not returnable. View return policy

Product Code. 16434497

Brand: Medchem Express HYB0806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Proguanil, an antimalarial prodrug, is metabolized to the active metabolite Cycloguanil (HY-12784). Proguanil is a dihydrofolate reductase (DHFR) inhibitor.

Proguanil, an antimalarial prodrug, is metabolized to the active metabolite Cycloguanil (HY-12784). Proguanil is a dihydrofolate reductase (DHFR) inhibitor.

Specifications

Chemical Name or Material Proguanil
Physical Form Solid
CAS 500-92-5
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C11H16ClN5
For Use With (Application) COVID-19-anti-virus
Solubility Information DMSO : ≥ 130 mg/mL (512.36 mM)
SMILES N=C(NC1=CC=C(Cl)C=C1)NC(NC(C)C)=N
Molecular Weight (g/mol) 253.73
Formula Weight 253.73
Percent Purity 98.81%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.