missing translation for 'onlineSavingsMsg'
Learn More

Proguanil hydrochloride, 97%

Product Code. 15658735 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15658735

Brand: Thermo Scientific Acros 461431000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 637-32-1 | C11H16ClN5·HCl | 290.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-32-1
Molecular Formula C11H16ClN5·HCl
Molecular Weight (g/mol) 290.19
InChI Key SARMGXPVOFNNNG-UHFFFAOYSA-N
Synonym proguanil hydrochloride, diguanyl, paludrine, proguanil hcl, chlorguanide hydrochloride, chloroquanil, bigumalum, chloroguanide hydrochloride, tirian hydrochloride, palusil hydrochloride
PubChem CID 9570076
IUPAC Name (1E)-1-[amino-(4-chloroanilino)methylidene]-2-propan-2-ylguanidine;hydrochloride
SMILES CC(C)N=C(N)N=C(N)NC1=CC=C(C=C1)Cl.Cl

Specifications

Melting Point 245.0°C to 250.0°C
Quantity 100 mg
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Formula Weight 290.19
Percent Purity 97%
Chemical Name or Material Proguanil hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.