missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Progranulin/PGRN Antibody Pair [HRP]

Product Code. 18250117 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18250117

Brand: Novus Biologicals™ NBP27928915PLATES

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody Pair

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Progranulin/PGRN.
TRUSTED_SUSTAINABILITY

Especificaciones

Assay Range 62.5 pg/mL
Assay Sensitivity 62.5-4000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Progranulin/PGRN.
Target Species Human
For Use With (Application) Sandwich ELISA
Gene Alias CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN
Gene ID (Entrez) 2896
Gene Symbol GRN
Host Species Mouse
Kit Contents
  • Detection Antibody: 0.1 mg/mL mouse anti-Progranulin/PGRN monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Capture Antibody: 0.5 mg/mL of mouse anti-Progranulin/PGRN monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 70 ng of recombinant Progranulin/PGRN.
  • Quantity 15 Plates
    Regulatory Status RUO
    Storage Requirements Storage is content dependent.
    Target Progranulin/PGRN
    Technique 1. Capture Antibody: Dilute to a working concentration of 2 μg/mL in CBS before coating. 2. Detection Antibody: Dilute to working concentration of 0.5 μg/mL in detection antibody dilution buffer before use. 3. Standard: Reconstitute with 1 ml detection antibody dilution buffer. After reconstitution, store at -20 to -80°C in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 4 ng/mL is recommended.
    Mostrar más Mostrar menos
    Corrección del contenido de un producto

    Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

    Título del producto

    Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

    Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado