missing translation for 'onlineSavingsMsg'
Learn More

Progranulin/PGRN Antibody (296605) [Janelia Fluor™ 525], Novus Biologicals™

Product Code. 30734486 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734486 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734486 Supplier Novus Biologicals Supplier No. FAB24201JF525

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Progranulin/PGRN Monoclonal antibody specifically detects Progranulin/PGRN in Human samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Progranulin/PGRN
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 296605
Conjugate Janelia Fluor 525
Formulation 50mM Sodium Borate
Gene Alias CLN11, GEP, GP88, granulin precursor, GRN, PCDGF, PEPI, PGRN
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Progranulin/PGRN, Met1-Leu593, Accession # P28799
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2896
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.