missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Progranulin Monoclonal Antibody (2D4-2F1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA1187
This item is not returnable.
View return policy
Description
By Western blot, MA1-187 detects Human Progranulin at ~85 kDa.
Progranulin is the precursor protein of Granulin also called as Proepithelin and PC cell-derived growth factor. Cleavage of the signal peptide produces mature Granulin. Both Granulin and smaller cleavage product of Granulin regulates cell growth. Progranulin stimulates proliferation of many epithelial cells, promotes their anchorage-independent growth and, when overexpressed, confers epithelial tumorigenicity. Progranulin/epithelin gene is multifunctional, with important constitutive roles in epithelial homeostasis, reproductive, immunological, and neuronal function. Progranulin constitutively expressed in a number of epithelia, particularly in the skin, GI tract, and reproductive system. Progranulin is expressed in immune cells in vivo and in specific neurons in the brain.
Specifications
| Progranulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| PBS with 1mg/mL BSA, 30% glycerol and 0.05% sodium azide | |
| P28799 | |
| GRN | |
| Recombinant human Progranulin. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2D4-2F1 | |
| Unconjugated | |
| GRN | |
| Acrogranin; acrogranulin; CLN11; epithelin; Epithelin precursor; Epithelin/granulin precursor; Epithelin-1; Epithelin-2; GEP; Glycoprotein of 88 Kda; GP88; granulin; Granulin A; Granulin B; Granulin C; Granulin D; Granulin E; Granulin F; Granulin G; Granulin precursor; Granulin-1; Granulin-2; Granulin-3; Granulin-4; Granulin-5; Granulin-6; Granulin-7; granulin-epithelin; granulins; GRN; Paragranulin; PC cell-derived growth factor; PCDGF; PEPI; Pgrn; Proepithelin; progranulin | |
| Mouse | |
| Protein A | |
| RUO | |
| 2896 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction