Learn More
Invitrogen™ Progesterone Receptor Monoclonal Antibody (PR-AT 4.14)

Description
MA1-410 detects the progesterone receptor (PR) from human, non-human primate, bovine, rat, rabbit, and mouse tissues. This antibody does not cross-react with androgen receptor, estrogen receptor or glucocorticoid receptor. MA1-410 has been successfully used in Western blot, immunohistochemical and immunoprecipitation procedures. By Western blot, this antibody detects a 94 kDa and a 120 kDa protein representing PRA and PRB, respectively from T47D whole cell lysate. MA1-410 performs better by western blot using BSA as the blocking agent and in antibody diluent. The MA1-410 immunogen is a synthetic peptide corresponding to the binding domain residues G(533) L P Q V Y P P Y L N Y L R P(547) of human Progesterone Receptor. MA1-410 can be used with blocking peptide PEP-016.
Specifications
Specifications
| Antigen | Progesterone Receptor |
| Applications | ChIP Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | PR-AT 4.14 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | PGR |
| Gene Accession No. | P06186, P06401, Q00175, Q63449 |
| Gene Alias | 9930019P03Rik; BB114106; ENSMUSG00000074510; LOW QUALITY PROTEIN: progesterone receptor; NR3C3; Nuclear receptor subfamily 3 group C member 3; p4 receptor; PGR; pot. alt. progesterone receptor; putative; PR; PR-A; PR-B; Progesterone receptor; progesterone receptor A; progesterone-receptor; putative form B; steroid hormone receptor; testicular progesterone receptor; unnamed protein product |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.