missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Progesterone Metabolites ELISA Kit

Product Code. 17194853
Change view
Click to view available options
Quantity:
96 Tests
Unit Size:
Pack of 96
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17194853 96 Tests Pack of 96
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17194853 Supplier Invitrogen™ Supplier No. EIAPROGM

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ELISA

A progesterone metabolites standard is provided to generate a standard curve for the assay and all samples are read off a user-generated standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with anti-rabbit IgG antibodies. Anti-rabbit progesterone metabolites antibodies and progesterone metabolites-peroxidase conjugate is added to the wells. The progesterone metabolites-peroxidase conjugate and any progesterone metabolites in the sample will compete to bind to the anti-rabbit progesterone metabolites antibodies. After incubation, the plate is washed and substrate is added. The substrate reacts with the bound progesterone metabolites-peroxidase conjugate. After a 30 min incubation, the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader at 450 nm. The intensity of the generated color corresponds inversely to the amount of progesterone metabolites present in the sample.

Progesterone, C21H30O2, also known as P4 (pregn-4-ene-3,20-dione) is a C-21 steroid hormone involved in the female menstrual cycle, gestation and embryogenesis of humans and other species. Progesterone belongs to a class of hormones called progestogens and is the major naturally occurring human progestogen. Progesterone is an essential regulator of human female reproductive function in the uterus, ovary, mammary gland and brain, and plays an important role in non-reproductive tissues such as the cardiovascular system, bone and the central nervous system.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number C21H30O2; Progesterone Metabolites; progestogen metabolites
Assay Range 102.4 to 10,000 pg/mL
Assay Sensitivity 51.2 pg/mL
Conjugate HRP
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
For Use With (Equipment) Colorimetric Microplate Reader
Interassay CV 7%
Intraassay CV 7
Kit Contents Anti-IgG coated 96 well plate, Standard, Assay Buffer Concentrate, Capture Antibody, HRP-conjugated Antigen, Wash Buffer Concentrate, TMB Substrate, Stop Solution, Plate Sealer
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 12.5 μL
Storage Requirements -20°C
Target Progesterone Metabolites
Test Time 45 mins
Total Assay Time 2 hrs 30 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.