missing translation for 'onlineSavingsMsg'
Learn More

Progesterone, MedChemExpress

Product Code. 16499076
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499076 100 mg 100mg
16471408 5 g 5g
16442529 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499076 Supplier Medchem Express Supplier No. HYN0437
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Progesterone is a steroid hormone that regulates the menstrual cycle and is crucial for pregnancy.

Progesterone is a steroid hormone that regulates the menstrual cycle and is crucial for pregnancy.

Specifications

Chemical Name or Material Progesterone
CAS 57-83-0
Color White
Purity Grade Notes Research
Molecular Formula C21H30O2
Quantity 100 mg
Synonym Pregn-4-ene-3,20-dione
Solubility Information DMSO : 25 mg/mL (79.50 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CC([C@H]1CC[C@@]2([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])CC[C@]12C)=O)=O
Molecular Weight (g/mol) 314.46
Formula Weight 314.46
Percent Purity 98.05%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.