Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Progesterone is a sex hormone required for suppression of estrogen signaling and is also involved in transcriptional activation
• Progesterone maintains the uterine endothelium and sustains pregnancy through suppression of muscle contractility
Applications
• Progesterone signaling is relevant to the study of breast cancer
Specifications
Specifications
| Melting Point | 126°C to 131°C |
| Color | Brown-Yellow to Yellow |
| Loss on Drying | 0.5% max. (500 mg, 105°C, 2 hrs) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Merck Index | 15, 7889 |
| Specific Rotation Condition | +190° (20°C c=1,ethanol) |
| Specific Rotation | + 190 |
| Show More |
Safety and Handling
- Progesterone
- Warning
- Carcinogenicity Category 2
- H351-Suspected of causing cancer.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.