missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ proFood™ 87-315 Series Blue Natural Rubber Latex Gloves

Product Code. 11712603 Shop All Ansell Products
Change view
Click to view available options
Size:
7
8
9
10
Unit Size:
12 pairs
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
11712603 8 12 pairs
11756744 7 12 pairs
12687021 9 12 pairs
11792633 10 12 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11712603 Supplier Ansell™ Supplier No. 87315080

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reusable latex gloves chlorinated outside to increase mechanical and chemical resistance.

  • Made from 100% pure latex

Applications:

  • Poultry processing
  • Beef and pig processing
  • Fish processing
  • Fruit and vegetable processing
TRUSTED_SUSTAINABILITY

Specifications

Back Material Rubber Latex
Cuff Material Natural Rubber Latex
Applications Industrial
Coating Material Natural Rubber Latex
Color Blue
Latex Free No
Material Rubber Latex
Product Type Latex Gloves
Size 8
Thickness (Metric) 0.4 mm
Glove Cuff Color Blue
Glove Cuff Style Pinked
Liner Material Cotton Flocking
Quantity 144 Pairs
Resistant To Liquid and mechanical resistant
Series 87-315
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.