missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Profilin 2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580892
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Profilin 2/PFN2.
Profilin 2 is a ubiquitous actin monomer-binding protein belonging to the profilin family. It is thought to regulate actin polymerization in response to extracellular signals.
Specifications
| Profilin 2 | |
| Polyclonal | |
| Unconjugated | |
| PFN2 | |
| D3S1319E; PFL; Pfn; PFN2; profilin 2; profilin II; Profilin-2 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 5217 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P35080 | |
| PFN2 | |
| Recombinant Human Profilin 2/PFN2 protein (Met1-Phe140). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction